missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPB8/HSP22 Rabbit anti-Human, Mouse, Rat, Clone: 3K10W1, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £405.00
Specifications
| Antigen | HSPB8/HSP22 |
|---|---|
| Clone | 3K10W1 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18374314
|
Novus Biologicals
NBP3-15464-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18360802
|
Bio-Techne
NBP3-15464-100UL |
100 μg |
£405.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSPB8/HSP22 Monoclonal antibody specifically detects HSPB8/HSP22 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| HSPB8/HSP22 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 26353 | |
| IgG | |
| Affinity purified |
| 3K10W1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Alpha-crystallin C chain, CRYAC, E2IG1E2-induced gene 1 protein, H11DHMN2, heat shock 22kDa protein 8, heat shock 27kDa protein 8, heat shock protein beta-8, HMN2, HMN2A, HSP22CMT2L, HspB8, Protein kinase H11, Small stress protein-like protein HSP22 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HSPB8/HSP22 (Q9UJY1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title