missing translation for 'onlineSavingsMsg'
Learn More

HSPB8/HSP22 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£305.00 - £435.00

Specifications

Antigen HSPB8/HSP22
Dilution Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20 - 1:50, Knockdown Validated
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18458090
View Documents
Novus Biologicals
NBP1-84945
0.1 mL
£435.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18429560
View Documents
Novus Biologicals
NBP1-84945-25ul
25 μL
£305.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HSPB8/HSP22 Polyclonal antibody specifically detects HSPB8/HSP22 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications

Specifications

HSPB8/HSP22
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown
Unconjugated
Rabbit
Protein Kinase
PBS (pH 7.2) and 40% Glycerol
26353
IgG
Immunogen affinity purified
Western Blot 0.04 - 0.4 ug/mL, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20 - 1:50, Knockdown Validated
Polyclonal
Purified
RUO
Human
Alpha-crystallin C chain, CRYAC, E2IG1E2-induced gene 1 protein, H11DHMN2, heat shock 22kDa protein 8, heat shock 27kDa protein 8, heat shock protein beta-8, HMN2, HMN2A, HSP22CMT2L, HspB8, Protein kinase H11, Small stress protein-like protein HSP22
This antibody was developed against Recombinant Protein corresponding to amino acids: LLDDGFGMDPFPDDLTASWPDWALPRLSSAWPGTLRSGMVPRGPTATARFGVPAEGRTPPPFPGEPWKVCVNVHSFKPEELMVKTKDGYVEVSGKHEEKQQEGGIVSKNFTKKIQLP
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.