missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPB7 Polyclonal Antibody

HSPB7 Polyclonal Antibody
Brand: Invitrogen PA569144
This item is not returnable.
View return policy
Description
HSPB7 Polyclonal Antibody for Western Blot
Specifications
| HSPB7 | |
| Polyclonal | |
| Unconjugated | |
| HSPB7 | |
| 27kDa; Cardiovascular heat shock protein; cvHsp; heat shock 27kD protein family member 7; heat shock 27kD protein family, member 7 (cardiovascular); heat shock 27kDa protein family member 7 (cardiovascular); heat shock 27kDa protein family, member 7 (cardiovascular); Heat shock protein; Heat shock protein 25 kDa 2; heat shock protein 25 kDa 2 (cardiovascular); heat shock protein B7; heat shock protein beta-7; heat shock protein family B (small) member 7; heat shock protein family, alpha-crystallin-related, b7; heat shock protein family, member 7 (cardiovascular); HSP; Hsp25-2; HspB7; Protein p19/6.8; zgc:103426 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29818 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P35385 | |
| HSPB7 | |
| synthetic peptide corresponding to a region of mouse HSPB7 (aa 120-169). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction