Learn More
Invitrogen™ HSPA9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579409
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human A549 whole cell, human Jurkat whole cell, human HepG2 whole cell, rat testis tissue, mouse testis tissue, mouse NIH/3T3 whole cell. IHC: human lung cancer tissue, rat liver tissue IHC-F: rat liver tissue. ICC/IF: A549 cell.
The HSP70 family is composed of four highly conserved proteins: HSP70, HSC70, GRP75 and GRP78. These proteins serve a variety of roles. GRP75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. GRP75 and GRP78 are unresponsive to heat stress and are induced by glucose deprivation.
Specifications
| HSPA9 | |
| Polyclonal | |
| Unconjugated | |
| HSPA9 | |
| 70 kDa heat shock protein; 74kDa; 75 kDa glucose-regulated protein; C3H-specific antigen; catecholamine-regulated protein 40; CRP40; CSA; epididymis secretory sperm binding protein Li 124m; EVPLS; GRP 75; GRP75; GRP-75; heat shock 70 kDa protein 9; heat shock 70 kDa protein 9B; heat shock 70kD protein 9B; heat shock 70kDa protein 9 (mortalin); heat shock 70kDa protein 9A; heat shock 70kDa protein 9B (mortalin-2); Heat shock protein; heat shock protein 9; heat shock protein 9A; heat shock protein cognate 74; heat shock protein family A (Hsp70) member 9; heat shock protein family A (Hsp70) member 9 S homeolog; heat shock protein, 74 kDa, A; heat shock protein, A; HEL-S-124m; Hsc74; HSP; Hsp74; Hsp74a; hspa9; hspa9.S; Hspa9a; hspa9-a; hspa9b; hspa9-b; hypothetical protein; I79_009139; MGC4500; mortalin; mortalin, perinuclear; mortalin2; mortalin-2; mot; MOT2; Mot-2; Mthsp70; mt-HSP70; MTHSP75; p66 MOT; p66-mortalin; pbp74; Peptide-binding protein 74; RCJMB04_2m8; SAAN; SIDBA4; stress-70 protein mitochondrial-like protein; stress-70 protein, mitochondrial; Stress-70 protein, mitochondrial-like protein; XELAEV_18020021mg | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15526, 291671, 3313 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| P38646, P38647, P48721 | |
| HSPA9 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human Grp75 (591-609aa DTETKMEEFKDQLPADECN). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.