missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13616-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody was raised against the C-terminal region of human HSPA6 and it may cross-react with HSPA1 (HSP70), HSPA1L and HSPA2.
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.Specifications
| HSPA6 | |
| Polyclonal | |
| Unconjugated | |
| HSPA6 | |
| Heat shock 70 kDa protein 6, Heat shock 70 kDa protein B, HSP70B, HSPA6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3310 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P17066 | |
| HSPA6 | |
| HSPA6 Fusion Protein Ag4522 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction