missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA5 Mouse anti-Human, HRP, Clone:1H11-1H7, Abnova™
Mouse monoclonal antibody raised against human HSPA5.
Brand: Abnova MAB17322.100ug
This item is not returnable.
View return policy
Description
When Chinese hamster K12 cells are starved of glucose, the synthesis of several proteins, called glucose-regulated proteins (GRPs), is markedly increased. Hendershot et al. (1994) [PubMed 8020977] pointed out that one of these, GRP78 (HSPA5), also referred to as ′immunoglobulin heavy chain-binding protein′ (BiP), is a member of the heat-shock protein-70 (HSP70) family and is involved in the folding and assembly of proteins in the endoplasmic reticulum (ER). Because so many ER proteins interact transiently with GRP78, it may play a key role in monitoring protein transport through the cell.[supplied by OMIM]Specifications
| HSPA5 | |
| Monoclonal | |
| HRP | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:2000) The optimal working dilution should be determined by the end user. |
|
| HSPA5 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1H11-1H7 | |
| heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa) | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| BIP, FLJ26106, GRP78, MIF2 | |
| His-tagged human HSPA5. | |
| 3309 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction