missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPA4L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558671
This item is not returnable.
View return policy
Description
Immunogen sequence: KCHAEHTPEE EIDHTGAKTK SAVSDKQDRL NQTLKKGKVK SIDLPIQSSL CRQLGQDLLN SY Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
HSPA4L possesses chaperone activity in vitro where it inhibits aggregation of citrate synthase.
Specifications
| HSPA4L | |
| Polyclonal | |
| Unconjugated | |
| HSPA4L | |
| 94kDa; AI461691; APG1; APG-1; heat shock 70 kDa protein 4L; heat shock 70 kDa protein 4-like protein; heat shock 70 kDa protein 4L-like; Heat shock 70 kDa protein 4L-like protein; heat shock 70kDa protein 4 like; heat shock 70kDa protein 4-like; heat shock 70-related protein APG-1; Heat shock protein; heat shock protein (hsp110 family); heat shock protein 4 like; heat shock protein 4-like; heat shock protein family A (Hsp70) member 4 like; HSP; Hsp4l; HSPA4L; HSPH3; LOW QUALITY PROTEIN: heat shock 70 kDa protein 4L; osmotic stress protein 94; OSP94; testis tissue sperm-binding protein Li 64n | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22824 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95757 | |
| HSPA4L | |
| Recombinant protein corresponding to Human HSPA4L. Recombinant protein control fragment (Product #RP-104170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction