missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA4L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544098
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: KCHAEHTPEE EIDHTGAKTK SAVSDKQDRL NQTLKKGKVK SIDLPIQSSL Sequence homology: Dog: 93%; Horse: 93%; Human: 100%; Mouse: 90%; Pig: 79%; Rat: 90%
HSPA4L Polyclonal antibody specifically detects HSPA4L in Human samples. It is validated for Western Blot.Specifications
HSPA4L | |
Polyclonal | |
Unconjugated | |
HSPA4L | |
94kDa; AI461691; APG1; APG-1; heat shock 70 kDa protein 4L; heat shock 70 kDa protein 4-like protein; heat shock 70 kDa protein 4L-like; Heat shock 70 kDa protein 4L-like protein; heat shock 70kDa protein 4 like; heat shock 70kDa protein 4-like; heat shock 70-related protein APG-1; Heat shock protein; heat shock protein (hsp110 family); heat shock protein 4 like; heat shock protein 4-like; heat shock protein family A (Hsp70) member 4 like; HSP; Hsp4l; HSPA4L; HSPH3; LOW QUALITY PROTEIN: heat shock 70 kDa protein 4L; osmotic stress protein 94; OSP94; testis tissue sperm-binding protein Li 64n | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22824 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O95757 | |
HSPA4L | |
Synthetic peptide directed towards the middle region of human HSPA4L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |