missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HspA4L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57695
This item is not returnable.
View return policy
Description
HspA4L Polyclonal antibody specifically detects HspA4L in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| HspA4L | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| APG1, APG-1, heat shock 70 kDa protein 4L, heat shock 70 kDa protein 4-like protein, heat shock 70kDa protein 4-like, Heat shock 70-related protein APG-1, heat shock protein (hsp110 family), Osmotic stress protein 94, Osp94 | |
| Rabbit | |
| 92 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Guinea pig: 92%; Mouse: 92%; Pig: 92%; Rabbit: 92%; Rat: 92%; Canine: 85%; Chicken: 84%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunoprecipitation | |
| O95757 | |
| HSPA4L | |
| Synthetic peptides corresponding to HSPA4L(heat shock 70kDa protein 4-like) The peptide sequence was selected from the C terminal of HSPA4L. Peptide sequence KDERYDHLDPTEMEKVEKCISDAMSWLNSKMNAQNKLSLTQDPVVKVSEI. | |
| Affinity purified | |
| RUO | |
| 22824 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction