missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPA4 Recombinant Rabbit Monoclonal Antibody (23GB6195), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552995
This item is not returnable.
View return policy
Description
The HSP 70 family is composed of four highly conserved proteins: HSP 70, HSC 70, GRP 75 and GRP 78. HSP 70 expression is strongly induced in response to heat stress. HSP 70 and HSC 70 play key roles in the cytosolic endoplasmic reticulum and mitochondrial import machinery and are found in both the cytosol and nucleus of mammalian cells. GRP 78 is localized in the endoplasmic reticulum, where it receives imported secretory proteins and is involved in the folding and translocation of nascent peptide chains. GRP 75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. It has been postulated that members of the HSP 70 family act as force-generating motors, relying on the hydrolysis of ATP for their activity.
Specifications
| HSPA4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| HSPA4 | |
| 70kDa; AI317151; APG2; APG-2; epididymis secretory sperm binding protein Li 5a; heat shock 70 kDa protein 4; heat shock 70kD protein 4; heat shock 70kDa protein 4; Heat shock 70-related protein APG-2; Heat shock protein; heat shock protein 4; heat shock protein Apg-2; heat shock protein family A (Hsp70) member 4; heat shock protein, 110 kDa; HEL-S-5a; HS24/P52; Hsp110; hsp70; hsp70 RY; Hsp70RY; Hspa4; HSPH2; Irp94; ischemia responsive 94 kDa protein; mKIAA4025; RY | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15525, 266759 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6195 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88600, Q61316 | |
| HSPA4 | |
| A synthesized peptide derived from human Hsp20 (90-160AA). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction