missing translation for 'onlineSavingsMsg'
Learn More

HspA1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen HspA1L
Dilution Western Blot 1:500 - 1:2000, ELISA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30230461
View Documents
Novus Biologicals
NBP3-35875-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30226756
View Documents
Novus Biologicals
NBP3-35875-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HspA1L Polyclonal antibody specifically detects HspA1L in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications

Specifications

HspA1L
ELISA, Western Blot
Unconjugated
Rabbit
Cancer
PBS (pH 7.3), 50% glycerol
3305
IgG
Affinity purified
Western Blot 1:500 - 1:2000, ELISA
Polyclonal
Purified
RUO
Human, Mouse
heat shock 10kDa protein 1-like, Heat shock 70 kDa protein 1-Hom, Heat shock 70 kDa protein 1L, heat shock 70 kDa protein 1-like, heat shock 70kD protein-like 1, heat shock 70kDa protein 1-like, HSP70-1L, HSP70-HOM, HSP70T, hum70t
Recombinant fusion protein containing a sequence corresponding to amino acids 342-641 of human HspA1L (NP_005518.3).,, Sequence:, STRIPKVQRLLQDYFNGRDLNKSINPDEAVAYGAAVQAAILMGDKSEKVQDLLLLDVAPLSLGLETAGGVMTALIKRNSTIPTKQTQIFTTYSDNQPGVLIQVYEGERAMTKDNNLLGRFDLTGIPPAPRGVPQIEVTFDIDANGILNVTATDKSTGKVNKITITNDKGRLSKEEIERMVLDAEKYKAEDEVQREKIAAKNALESYAFNMKSVVSDEGLKGKISESDKNKILDKCNELLSWLEVNQLAEKDEFDHKRKELEQMCNPIITKLYQGGCTGPACGTGYVPGRPATGPTIEEVD
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.