missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSPA1B Recombinant Rabbit Monoclonal Antibody (K01_2U22)
GREENER_CHOICE

Product Code. 30828601
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828601 100 μL
30825850 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828601 Supplier Invitrogen™ Supplier No. MA564278
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

HSPA1B Recombinant Monoclonal Antibody for IHC (P), Western Blot

This intronless gene encodes a 70 kDa heat shock protein which is a member of the heat shock protein 70 family. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2U22
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Hspa1b
Gene Accession No. P0DMV9, P0DMW1, P17879
Gene Alias 68 kDa heat shock protein; Hcp70.1; Heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; Heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; heat shock 70kD protein 1B; heat shock 70kD protein 1B (mapped); heat shock 70kDa protein 1B; Heat shock protein; heat shock protein 1B; heat shock protein 70.1; heat shock protein family A (Hsp70) member 1B; heat shock protein, 70 kDa 1; HSP; hsp68; Hsp70; HSP70.1; HSP70.1/HSP70.2; HSP70.2; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; HSP70-1B; HSP70-2; Hsp70a1; HSP70I; Hsp72; HSPA1; Hspa1a; HSPA1B; Hspa2; HSX70
Gene Symbols Hspa1b
Host Species Rabbit
Immunogen Recombinant protein of mouse Hsp70.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15511, 294254, 3304
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.