missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSPA1B Polyclonal Antibody

Rabbit Polyclonal Antibody
£133.00 - £397.00
Specifications
| Antigen | HSPA1B |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17770584
![]() |
Invitrogen™
PA5119176 |
200 μL |
£397.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17733294
|
Invitrogen™
PA5121895 |
20 μL |
£133.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSPA1B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This intronless gene encodes a 70 kDa heat shock protein which is a member of the heat shock protein 70 family. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.Specifications
| HSPA1B | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| Hspa1b | |
| 68 kDa heat shock protein; Hcp70.1; Heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; Heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; heat shock 70kD protein 1B; heat shock 70kD protein 1B (mapped); heat shock 70kDa protein 1B; Heat shock protein; heat shock protein 1B; heat shock protein 70.1; heat shock protein family A (Hsp70) member 1B; heat shock protein, 70 kDa 1; HSP; hsp68; Hsp70; HSP70.1; HSP70.1/HSP70.2; HSP70.2; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; HSP70-1B; HSP70-2; Hsp70a1; HSP70I; Hsp72; HSPA1; Hspa1a; HSPA1B; Hspa2; HSX70 | |
| Hspa1b | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P0DMV9, P17879 | |
| 15511, 3304 | |
| Recombinant protein Heat Shock 70kDa Protein 1B. The antigen corresponds to amino acid range 1-642 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title