missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HspA1B Monoclonal antibody specifically detects HspA1B in Bovine,Human,Mouse,Porcine,Rat samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence,ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin),Proximity Ligation Assay,Dot Blot
Specifications
Specifications
| Antigen | HspA1B |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Proximity Ligation Assay, Dot Blot |
| Classification | Monoclonal |
| Clone | 3B7 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Proximity Ligation Assay, Dot Blot |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_005336 |
| Gene Alias | FLJ54328, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1B, heat shock 70kDa protein 1B, HSP70.1/HSP70.2, HSP70-1/HSP70-2, HSP70-1B, HSP70-2, HSPA1, HSPA1A |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?