missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSPA1A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534772
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, NIH-3T3, Mouse brain, PC-12, Rat brain, Jurkat, Raji, 293T, A431, HeLa, HepG2, H1299, HCT116, MCF-7, NT2D1, PC-3, U87-MG, Mesenchymal stem cells. Predicted reactivity: Mouse (97%), Rat (98%), Xenopus laevis (85%), Sheep (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.Specifications
HSPA1A | |
Polyclonal | |
Unconjugated | |
HSPA1A | |
68 kDa heat shock protein; APG2; APG-2; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; epididymis secretory sperm binding protein Li 5a; heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70 kDa protein 4; heat shock 70kD protein 1A; heat shock 70kD protein 4; heat shock 70kDa protein 1A; heat shock 70kDa protein 4; Heat shock 70-related protein APG-2; heat shock protein 1A; heat shock protein 70; heat shock protein 70-1; heat shock protein Apg-2; heat shock protein family A (Hsp70) member 1A; heat shock protein family A (Hsp70) member 4; heat shock protein, 110 kDa; heat shock protein, 70 kDa 3; heat shock-induced protein; HEL-S-103; HEL-S-5a; HS24/P52; hsp68; hsp70; hsp70 RY; HSP70.1; HSP70.1/2; HSP70.1/HSP70.2; Hsp70.3; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; Hsp70-2; Hsp70-3; Hsp70A1; HSP7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
193740, 24472, 3303 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.62 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P0DMV8, P0DMW0, Q61696 | |
HSPA1A | |
Recombinant protein fragment contains a sequence corresponding to a region within amino acids 308 and 569 of HSP70 1A | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |