missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSP90 beta Recombinant Rabbit Monoclonal Antibody (23GB2785), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553485
This item is not returnable.
View return policy
Description
Heat shock proteins (HSP) are expressed in response to various biological stresses, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, aryl hydrocarbon (Ah) receptor, and some kinases.
Specifications
| HSP90 beta | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Hsp90ab1 | |
| 90kDa; 90-kDa heat shock protein beta; AL022974; C81438; D6S182; heat shock 84 kDa; heat shock 90 kDa protein; heat shock 90kD protein 1, beta; heat shock 90kDa protein 1, beta; heat shock cognate protein HSP 90-beta; Heat shock protein; heat shock protein 1, beta; heat shock protein 90 alpha (cytosolic), class B member 1; heat shock protein 90 alpha family class B member 1; heat shock protein 90 beta; heat shock protein 90 kDa; heat shock protein 90 kDa alpha class B member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein HSP 90-beta; heat shock protein, 84 kDa 1; HSP; HSP 84; HSP 90; Hsp84; Hsp84-1; Hsp90; hsp90 beta; HSP90AB1; HSP90B; HSP90-beta; HSPC2; HSPCB; retinal degeneration slow protein; RP1-302G2.1; TSTA; Tumor-specific transplantation 84 kDa antigen; Unknown (protein for IMAGE:7943063) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15516, 301252, 3326 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2785 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P08238, P11499, P34058 | |
| Hsp90ab1 | |
| A synthesized peptide derived from human Hsp90 beta (500-724AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction