missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HSP90 beta Monoclonal Antibody (5G4D2)

Product Code. 12867882
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12867882 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12867882 Supplier Invitrogen™ Supplier No. MA515863

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-15863 targets HSP90 beta in WB, IF, IHC, and Flow applications and shows reactivity with Human, Non-human primate, mouse and rat samples.

Heat shock proteins (HSP) are expressed in response to various biological stresses, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, aryl hydrocarbon (Ah) receptor, and some kinases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP90 beta
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5G4D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Hsp90ab1
Gene Accession No. P08238, P11499, P34058
Gene Alias 90kDa; 90-kDa heat shock protein beta; AL022974; C81438; D6S182; heat shock 84 kDa; heat shock 90 kDa protein; heat shock 90kD protein 1, beta; heat shock 90kDa protein 1, beta; heat shock cognate protein HSP 90-beta; Heat shock protein; heat shock protein 1, beta; heat shock protein 90 alpha (cytosolic), class B member 1; heat shock protein 90 alpha family class B member 1; heat shock protein 90 beta; heat shock protein 90 kDa; heat shock protein 90 kDa alpha class B member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein HSP 90-beta; heat shock protein, 84 kDa 1; HSP; HSP 84; HSP 90; Hsp84; Hsp84-1; Hsp90; hsp90 beta; HSP90AB1; HSP90B; HSP90-beta; HSPC2; HSPCB; retinal degeneration slow protein; RP1-302G2.1; TSTA; Tumor-specific transplantation 84 kDa antigen; Unknown (protein for IMAGE:7943063)
Gene Symbols Hsp90ab1
Host Species Mouse
Immunogen Purified recombinant fragment of human HSP90AB1 expressed in E. Coli.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15516, 301252, 3326
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.