missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55509
This item is not returnable.
View return policy
Description
HSP90 beta Polyclonal specifically detects HSP90 beta in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications
| HSP90AB1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Knockout Validated, KnockDown Validated | |
| D6S182, Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, Heat Shock Protein 90 Alpha Family Class B Member 1, Heat Shock Protein 90 KDa, heat shock protein 90kDa alpha (cytosolic), class B member 1, Heat Shock Protein 90kDa Alpha Family Class B Member 1, heat shock protein HSP 90-beta, HSP 84, HSP 90, HSP84, HSP90B, HSP90-BETA, HSPC2, HSPCB | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| HSP90AB1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DEDEVAAEEPNAAVPDEIPPLEGDEDASR | |
| 100 μL | |
| Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones | |
| 3326 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction