missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 Antibody (AC88), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NB100-1972-0.025MG
This item is not returnable.
View return policy
Description
HSP90 Monoclonal specifically detects HSP90 in Human, Mouse, Rat, Porcine, Bacteria, Canine, Chicken, Fish, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications
| HSP90 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 ug/ml, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular) | |
| FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| Mouse | |
| 90 kDa | |
| 0.025 mg | |
| Primary | |
| Recognizes both Hsp90 alpha and beta. It reacts with the rodent glucocorticoid receptor and with proliferation potential proteins (P2PS) with apparent molecular masses of 30-40 kDa. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Flow Cytometry | |
| AC88 | |
| Unconjugated | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| HSP90AA1 | |
| Achlya ambisexualis (water mold) Hsp90. | |
| Protein G purified | |
| RUO | |
| 3320 | |
| Rabbit, Sheep, Rat, Primate, Mouse, Bacteria, Fish, Chicken, Human, Canine, Pig, Guinea Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction