missing translation for 'onlineSavingsMsg'
Learn More

HSP90 Antibody (341320) [Biotin], Novus Biologicals™

Product Code. 30734717 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734717 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734717 Supplier Novus Biologicals Supplier No. FAB3286B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSP90 Monoclonal antibody specifically detects HSP90 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP90
Applications Western Blot
Classification Monoclonal
Clone 341320
Conjugate Biotin
Formulation PBS
Gene Alias FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
Host Species Mouse
Immunogen E. coli-derived recombinant human HSP90β, Met1-Asp724, Accession # P08238
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3320
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.