missing translation for 'onlineSavingsMsg'
Learn More

HSP90 alpha Monoclonal Antibody (3G3), Invitrogen™

Product Code. 11515732
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11515732 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11515732 Supplier Invitrogen Supplier No. MA3011

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA3-011 detects heat shock protein 90 kDa (HSP90) from human, mouse, rat, rainbow trout, and chicken tissues. MA3-011 has been successfully used in Western blot, immunofluorescence, immunohistochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects a 90 kDa protein representing HSP90 from Hepa 1 cell extract. It has been reported that up to 90% of cytosolic HSP90 can be immunoprecipitated with this product under low ionic strength conditions. For Western blots using MA3-011, nitrocellulose is recommended. PVDF membranes typically do not yield acceptable results with this antibody. Before use, centrifuge suspension (4000 g for 15 minutes) and discard supernatant. Resuspend pellet in 200 μL of PBS, pH 7.2. If desired, 0.05% sodium azide may be added to resuspended solution. Removal of residual ammonium sulfate is not usually necessary for immunoprecipitation and Western blot procedures, however, any other uses may require its removal by dialysis or equivalent method. Stable for 6 months after PBS resuspension. The MA3-011 antigen is purified HSP90 from Hepa 1 cytosol.

Heat shock proteins (HSP) are proteins that are expressed in response to various biological stress, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, and some kinases. It has been suggested that HSP90 is necessary to maintain structural integrity of at least some steroid receptors to allow for ligand binding during receptor activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP90 alpha
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3G3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with Ammonium sulfate and no preservative
Gene Hsp90aa1
Gene Accession No. P07900, P07901, P11501, P82995
Gene Alias 86kDa; 89kDa; 90-kDa heat shock protein; 90-kDa heat shock protein alpha; akineto; AL024080; AL024147; EL52; epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; fb17b01; heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 86; heat shock protein 90; heat shock protein 90 alpha family class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1, tandem duplicate 1; heat shock protein 90-alpha 1; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein alpha; Heat shock protein HSP 90-alpha; Heat shock protein HSP 90-alpha 1; heat shock protein HSP90 alpha; heat shock protein hsp90alpha; heat shock protein, 1; heat shock protein, 86 kDa 1; heat shock protein, 89 kDa; Heat shock protein90; HEL-S-65p; HSP; HSP 86; HSP 90 alpha; hsp4; HSP86; Hsp86-1; Hsp89; HSP89A; HSP90; hsp90a; hsp90a protein; hsp90a.1; hsp90aa1; hsp90aa1.1; hsp90alpha; HSP90N; HSPC1; Hspca; HSPCA protein; HSPCAL1; HSPCAL4; HSPN; hypothetical protein; LAP2; LAP-2; Lipopolysaccharide-associated protein 2; LPS-associated protein 2; renal carcinoma antigen NY-REN-38; slo; sloth; TSTA; Tumor-specific transplantation 86 kDa antigen; wu:fb17b01; zgc:86652
Gene Symbols Hsp90aa1
Host Species Mouse
Immunogen Purified mouse HSP90.
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15519, 299331, 3320, 423463
Target Species Chicken, Fish, Human, Mouse, Non-human Primate, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.