Learn More
Invitrogen™ HSP90 alpha Human ELISA Kit
For the quantitative detection of human HSP90 alpha.
£437.00 - £2790.00
Specifications
Accession Number | P07900 |
---|---|
Assay Range | 0.156-10 ng/mL |
Assay Sensitivity | 0.03 ng/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15571617
|
Invitrogen™
BMS2090TEN |
10 x 96 Tests |
£2790.00
Pack of 10 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15561617
|
Invitrogen™
BMS2090 |
96 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The Human Heat Shock Protein 90 alpha (Hu HSP90 alpha) ELISA quantitates Hu HSP90 alpha in human serum, plasma cell lysate. The assay will exclusively recognize both natural and recombinant Hu HSP90 alpha. Principle of the method The Human HSP90 alpha solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
P07900 | |
0.03 ng/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
10% | |
Pre-coated 96 well plate, Standard, Standard, Sample Diluent, Assay Buffer concentrate, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Controls, Controls | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. |
0.156-10 ng/mL | |
Biotin | |
Cell Lysates, Plasma, Serum | |
ELISA | |
3320 | |
8.6% | |
HRP | |
Cell Lysate,100 μL; Plasma, 10 μL; Serum, 10 μL | |
Heat Shock Protein 90 alpha | |
1 hr. 20 min. |