missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 alpha Antibody (Hyb-K41009), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-61641
This item is not returnable.
View return policy
Description
HSP90 alpha Monoclonal specifically detects HSP90 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HSP90 alpha | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects 90kD proteins corresponding to the molecular mass of hsp90alpha. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Hyb-K41009 | |
| Unconjugated | |
| P07900 | |
| HSP90AA1 | |
| Recombinant human HSP90alpha; Specificity mapped to amino acids 604-731 | |
| 0.1 mg | |
| Cancer, Hypoxia, Membrane Trafficking and Chaperones | |
| 3320 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction