missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 alpha Antibody (HSP90AA1/7247) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26954JF635
This item is not returnable.
View return policy
Description
HSP90 alpha Monoclonal antibody specifically detects HSP90 alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HSP90 alpha | |
| Monoclonal | |
| Janelia Fluor 635 | |
| FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| Recombinant fragment (around aa500-700) of human HSP90 alpha protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones | |
| 3320 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| HSP90AA1/7247 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction