missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP70 Rat anti-Fruit fly, Clone:7FB, Abnova™
Rat monoclonal antibody raised against drosophila HSP70.
Brand: Abnova MAB16783.100ug
This item is not returnable.
View return policy
Specifications
| HSP70 | |
| Monoclonal | |
| Unconjugated | |
| ELISA Immunocytochemistry Immunofluorescence Western Blot (1:2000) The optimal working dilution should be determined by the end user. |
|
| Hsp70Bb | |
| Rat | |
| 100 μg | |
| Fruitfly | |
| Antibody | |
| IgG2b |
| ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 7FB | |
| Heat-shock-protein-70Bb | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| CG31359, DMHSP7D1, HSP70, Hsp 70, Hsp70(87C), Hsp70A7d, Hsp70B, Hsp70Bbc, Hsp70Bc, dhsp70, hsp-70, hsp70 87C, unnamed | |
| Drosophila tissue culture cells. | |
| 48582 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction