missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25405-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody recognizes two isoforms of HSP70 around 70 kDa and 60 kDa, respectively.
This intronless gene encodes a 70 kDa heat shock protein which is a member of the heat shock protein 70 family. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.Specifications
HSP70 | |
Polyclonal | |
Unconjugated | |
HSPA1B | |
Heat shock 70 kDa protein 1/2, HSP70, HSPA1, HSPA1A, HSPA1B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15511, 294254, 3304 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P0DMV9, P0DMW1, P17879 | |
HSPA1B | |
HSP70 Fusion Protein Ag13700 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |