missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP70 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25405-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizes two isoforms of HSP70 around 70 kDa and 60 kDa, respectively.
This intronless gene encodes a 70 kDa heat shock protein which is a member of the heat shock protein 70 family. In conjunction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.Specifications
| HSP70 | |
| Polyclonal | |
| Unconjugated | |
| HSPA1B | |
| Heat shock 70 kDa protein 1/2, HSP70, HSPA1, HSPA1A, HSPA1B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15511, 294254, 3304 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P0DMV9, P0DMW1, P17879 | |
| HSPA1B | |
| HSP70 Fusion Protein Ag13700 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion