missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp70 interacting protein HIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16894
This item is not returnable.
View return policy
Description
Hsp70 interacting protein HIP Polyclonal specifically detects Hsp70 interacting protein HIP in Human samples. It is validated for Western Blot.
Specifications
| Hsp70 interacting protein HIP | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| AAG2, aging-associated protein 2, FAM10A1FAM10A4, heat shock 70kD protein binding protein, Hip, HIPFLJ27260, HOP, hsc70-interacting protein, Hsp70-interacting protein, HSPABP1, P48MGC129952, PRO0786, Progesterone receptor-associated p48 protein, Protein FAM10A1, Putative tumor suppressor ST13, Renal carcinoma antigen NY-REN-33, SNC6HSPABP, suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein), suppression of tumorigenicity 13 (colon carcinoma) (Hsp70-interacting protein), Suppression of tumorigenicity 13 protein | |
| Rabbit | |
| 41 kDa | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones | |
| 6767 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:5000-1:20000 | |
| P50502 | |
| ST13 | |
| Recombinant protein encompassing a sequence within the center region of human HIP. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Rat (95%), Bovine (97%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction