missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp70 interacting protein HIP Antibody (CL3708) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61615MFV500
This item is not returnable.
View return policy
Description
Hsp70 interacting protein HIP Monoclonal antibody specifically detects Hsp70 interacting protein HIP in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Hsp70 interacting protein HIP | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| AAG2, aging-associated protein 2, FAM10A1FAM10A4, heat shock 70kD protein binding protein, Hip, HIPFLJ27260, HOP, hsc70-interacting protein, Hsp70-interacting protein, HSPABP1, P48MGC129952, PRO0786, Progesterone receptor-associated p48 protein, Protein FAM10A1, Putative tumor suppressor ST13, Renal carcinoma antigen NY-REN-33, SNC6HSPABP, suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein), suppression of tumorigenicity 13 (colon carcinoma) (Hsp70-interacting protein), Suppression of tumorigenicity 13 protein | |
| This antibody was developed using a recombinant protein derived from P50502, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Lipid and Metabolism, Membrane Trafficking and Chaperones | |
| 6767 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL3708 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction