missing translation for 'onlineSavingsMsg'
Learn More

HSP70/HSPA1A Antibody (OTI3G9), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18402873 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402873 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402873 Supplier Novus Biologicals Supplier No. NBP270954MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSP70/HSPA1A Monoclonal antibody specifically detects HSP70/HSPA1A in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP70/HSPA1A
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3G9
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B
Host Species Mouse
Immunogen Full length human recombinant protein of human HSPA1A (NP_005336) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3303
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.