missing translation for 'onlineSavingsMsg'
Learn More

HSP70/HSPA1A Antibody, DyLight 350, Novus Biologicals™

Product Code. 18373901 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373901 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373901 Supplier Novus Biologicals Supplier No. NBP177455UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSP70/HSPA1A Polyclonal specifically detects HSP70/HSPA1A in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP70/HSPA1A
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B
Gene Symbols HSPA1A
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Hsp70 protein (between residues 550-600) [Uniprot: P08107]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3303.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.