missing translation for 'onlineSavingsMsg'
Learn More

HSP70/HSPA1A Antibody (5A5), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications

£164.00

Specifications

Antigen HSP70/HSPA1A
Clone 5A5
Concentration 1 mg/mL
Dilution Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1ug, Immunohistochemistry-Paraffin 1:10-1:500, Microarray
Applications Western Blot
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18586091
View Documents
Novus Biologicals
NBP1-21690-0.025mg
0.025 mg
£164.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HSP70/HSPA1A Monoclonal specifically detects HSP70/HSPA1A in Human, Mouse, Rat, Amphibian, Chicken, Drosophila, Fish, Yeast samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications

Specifications

HSP70/HSPA1A
1 mg/mL
Western Blot
Unconjugated
Mouse
Human
dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B
HSPA1A
IgG1
Protein G purified
Detects approx 70kDa. May detect HSP70, HSC70, Grp78 and HSP72.
5A5
Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1ug, Immunohistochemistry-Paraffin 1:10-1:500, Microarray
Monoclonal
Purified
RUO
P08107
3303
Human recombinant HSP70 overexpressed in E.coli
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.