missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP70/HSPA1A Antibody (1H11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59339-25ug
This item is not returnable.
View return policy
Description
HSP70/HSPA1A Monoclonal specifically detects HSP70/HSPA1A in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Flow (Cell Surface).
Specifications
| HSP70/HSPA1A | |
| Monoclonal | |
| 0.5 mg/ml | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Binds the altered conformation of HSP70 embedded in the cell membrane and to intracellular HSP70 to a lesser extent. | |
| Store at -20°C. | |
| IgG1 |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1H11 | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| HSPA1A | |
| Human native HSP70 protein | |
| 25 μg | |
| Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
| 3303 | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction