missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP70/HSPA1A, ac Lys77 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-61122
This item is not returnable.
View return policy
Description
HSP70/HSPA1A Polyclonal specifically detects HSP70/HSPA1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative).
Specifications
| HSP70/HSPA1A (ac Lys77) | |
| Polyclonal | |
| Western Blot 1:1000, ELISA 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:60, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative) 1:50 | |
| dnaK-type molecular chaperone HSP70-1, FLJ54303, FLJ54370, FLJ54392, FLJ54408, FLJ75127, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1A, heat shock 70kDa protein 1A, heat shock-induced protein, HSP70.1/HSP70.2, HSP70-1, HSP70-1/HSP70-2, HSP70-1A, HSP70I, HSP72, HSPA1, HSPA1B | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Cellular Markers, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
| 3303 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS pH 7.4 and 50% glycerol with 0.09% Sodium Azide | |
| HSPA1A | |
| Synthetic peptide from the N-terminal of Human HSP70 Acetyl Lys77 (1-100 aa), conjugated to Keyhole Limpet Haemocyanin (KLH). | |
| 100 μL | |
| Primary | |
| Binds to acetylated lysine 77 on HSP70. Detects 70 kDa band. | |
| Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction