missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep, Clone: HSPD1/6496R, Novus Biologicals™
Rabbit Monoclonal Antibody
£235.00
Specifications
| Antigen | HSP60 |
|---|---|
| Clone | HSPD1/6496R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
HSP60 Monoclonal specifically detects HSP60 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Spécification
| HSP60 | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
| 10 mM PBS with 0.05% BSA | |
| 3329 | |
| IgG κ | |
| Protein A or G purified |
| HSPD1/6496R | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
| Recombinant human full-length HSP60 protein | |
| Primary | |
| Store at 4°C. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit