missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Mouse anti-Human, Mouse, Rat, Clone: 2F10E7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66041-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.Specifications
| HSP60 | |
| Monoclonal | |
| 0.95 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P10809, P63038, P63039 | |
| HSPD1 | |
| HSP60 Fusion Protein Ag7848 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
| 2F10E7 | |
| Unconjugated | |
| HSPD1 | |
| Heat shock protein 60, HSP 60, HSP60, HSP65, HSPD1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 15510, 3329, 63868 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction