missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSP60 Monoclonal Antibody (LK2), FITC
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545113
This item is not returnable.
View return policy
Description
0.25 μg/mL of MA5-45113 was sufficient for detection of HSP60 in 10 μg of heat shocked HeLa cell lysate by colorimetric immunoblot analysis using goat anti-mouse IgG as the secondary antibody. Detects approximately 60kDa.
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.
Specifications
| HSP60 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative | |
| P10809, P18687, P19882, P31081, P49818, P63038, P63039, Q5ZL72 | |
| HSP60, HSPD1 | |
| Recombinant human HSP60. | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Chicken, Fish, Fish, Bacteria, Guinea Pig, Nematode, Plant, Yeast | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot | |
| LK2 | |
| FITC | |
| HSPD1 | |
| 60 kDa chaperonin; 60 kDa heat shock protein; 60 kDa heat shock protein, mitochondrial; 60kDa; Chaperonin 60; CPN60; GROEL; heat shock 60kD protein 1 (chaperonin); heat shock 60kDa protein 1; heat shock 60kDa protein 1 (chaperonin); Heat shock protein; heat shock protein 1 (chaperonin); heat shock protein 60; heat shock protein 60 (liver); heat shock protein 65; heat shock protein family D (Hsp60) member 1; heat shock protein, 60 kDa; HLD4; HSP; Hsp60; HSP-60; Hsp60s1; HSP65; HSP-65; Hspd1; Hspd1-30p; HuCHA60; hypothetical protein; I79_011398; mitochondrial matrix protein P1; P1 protein; P60 lymphocyte protein; RCJMB04_7g5; short heat shock protein 60 Hsp60s1; SPG13; Unknown (protein for IMAGE:7943409) | |
| Mouse | |
| Protein G | |
| RUO | |
| 100349912, 100689473, 100715481, 15510, 3329, 424059, 478854, 492279, 511913, 63868, 850963 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction