missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£240.00 - £386.00
Specifications
| Antigen | HSP60 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18201424
|
Novus Biologicals
NBP2-55503 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645406
|
Novus Biologicals
NBP2-55503-25ul |
25 μL |
£240.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSP60 Polyclonal specifically detects HSP60 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| HSP60 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 3329 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KGVITVKDGKTLNDELEIIEGMKFDRGYISPYFINTSKGQKCEFQDAYVLLSEKKISSIQSIVPALEIANAHRKPLVIIAEDVDGEALSTLVLNRLKVGLQVVAVKAPGFGDNRKNQLKDMAIATGGAVFGEEGL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
| HSPD1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title