missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Antibody (HSPD1/780), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47843IR
This item is not returnable.
View return policy
Description
HSP60 Monoclonal antibody specifically detects HSP60 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array, Immunofluorescence, CyTOF
Specifications
| HSP60 | |
| Monoclonal | |
| DyLight 755 | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
| Recombinant human HSP60 protein (Uniprot: P10809) | |
| 0.1 mL | |
| Apoptosis, Cellular Markers, Loading Controls, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
| 3329 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HSPD1/780 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction