missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Antibody (4B9/89) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22440-0.025mg
This item is not returnable.
View return policy
Description
HSP60 Monoclonal specifically detects HSP60 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| HSP60 | |
| Monoclonal | |
| Unconjugated | |
| P10809 | |
| HSPD1 | |
| Human placental HSP60. | |
| 0.025 mg | |
| Apoptosis, Cellular Markers, Membrane Trafficking and Chaperones, Mitochondrial Markers | |
| 3329 | |
| Reconstitute with PBS. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 4B9/89 | |
| Western Blot 1:100 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:200 | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody reacts with Human, Rat and Primate. | |
| Human, Mouse, Rat, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction