missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Antibody (2E1/53), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-5479
This item is not returnable.
View return policy
Description
HSP60 Monoclonal specifically detects HSP60 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| HSP60 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 - 10 ug/ml, ELISA 1 - 10 ug/ml, Immunohistochemistry 1 - 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 ug, Immunohistochemistry-Paraffin 1 - 10 ug/ml | |
| HSPD1 | |
| Human placental HSP60. | |
| 100 μg | |
| Apoptosis, Membrane Trafficking and Chaperones | |
| 3329 | |
| Reconstitute with PBS. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 2E1/53 | |
| Unconjugated | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant) | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Detects Hsp 60 from human cells, tissues and recombinant protein preparations. This displays no other protein or species cross-reactivity. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction