missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse HSP47 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£171.00 - £377.00

Specifications

Antigen Hsp47
Clone 950806
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-25 ug/mL
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18711708
GREENER_CHOICE
View Documents
R&D Systems
MAB9166-SP
25 μg
£171.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18780749
GREENER_CHOICE
View Documents
R&D Systems
MAB9166-100
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hsp47 Monoclonal specifically detects Hsp47 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Hsp47
Western Blot 0.1 ug/mL, Immunohistochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
(collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, BERF-1, CBP1, CBP2, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), OI10, PPROM, proliferation-inducing gene 14, RA-A47, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH1, SERPINH2colligin
SERPINH1
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
950806
Western Blot, Immunohistochemistry
Unconjugated
Mouse
Human, Mouse
P50454
871
Chinese hamster ovary cell line CHO-derived recombinant human HSP47 Ala19-Asp412 Accession # P50454
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human and mouse HSP47 in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.