missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsp47 Antibody (M16.10A1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 14 publications
£150.00 - £599.00
Specifications
| Antigen | Hsp47 |
|---|---|
| Clone | M16.10A1 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry, Immunofluorescence, Western Blot, Flow Cytometry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18077223
|
Novus Biologicals
NBP1-97491-0.025MG |
0.025 mg |
£150.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18708873
|
Novus Biologicals
NBP1-97491-0.05mg |
0.05 mg |
£376.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18728393
|
Novus Biologicals
NBP1-97491-0.2mg |
0.2 mg |
£599.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hsp47 Monoclonal specifically detects Hsp47 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.Specifications
| Hsp47 | |
| 1.0 mg/mL | |
| Immunohistochemistry, Immunofluorescence, Western Blot, Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 871 | |
| Native rat HSP47. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| M16.10A1 | |
| Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Canine, Rat, Bovine, Sheep, Chicken, Human, Guinea Pig, Hamster, Mouse, Rabbit, Monkey, Pig | |
| (collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin | |
| SERPINH1 | |
| IgG2b | |
| Protein G purified | |
| Recognizes Sheep HSP47. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title