missing translation for 'onlineSavingsMsg'
Learn More

Hsp47 Antibody (M16.10A1), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18419295 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419295 0.2 mL 0.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419295 Supplier Novus Biologicals Supplier No. NBP197491AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hsp47 Monoclonal antibody specifically detects Hsp47 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp47
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone M16.10A1
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias (collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin
Host Species Mouse
Immunogen Native rat HSP47.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 871
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.