missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Hsp47 Monoclonal antibody specifically detects Hsp47 in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | Hsp47 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1C4-1A6 |
| Conjugate | FITC |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | PBS |
| Gene Alias | (collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin |
| Host Species | Mouse |
| Immunogen | Human HSP47, full length |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?