missing translation for 'onlineSavingsMsg'
Learn More

Hsp47 Antibody (1C4-1A6), DyLight 550, Novus Biologicals™

Product Code. 18407076 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407076 0.2 mL 0.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407076 Supplier Novus Biologicals Supplier No. NBP222400R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hsp47 Monoclonal antibody specifically detects Hsp47 in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp47
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 1C4-1A6
Conjugate DyLight 550
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias (collagen binding protein 1), Arsenic-transactivated protein 3, AsTP3, CBP1, CBP2RA-A47, Cell proliferation-inducing gene 14 protein, Collagen-binding protein, colligen, Colligin, colligin-1, colligin-2, gp46, HSP47serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1), member 2, member 2, (collagen-binding protein 2), PPROM, proliferation-inducing gene 14, rheumatoid arthritis antigen A-47,47 kDa heat shock protein, Rheumatoid arthritis-related antigen RA-A47, serine (or cysteine) proteinase inhibitor, clade H (heat shock protein 47), serpin H1, serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, SERPINH2colligin
Host Species Mouse
Immunogen Human HSP47, full length
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 871
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.