missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP40/DNAJB1 Antibody (OTI1F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-03433
This item is not returnable.
View return policy
Description
HSP40/DNAJB1 Monoclonal specifically detects HSP40/DNAJB1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HSP40/DNAJB1 | |
| Monoclonal | |
| 1.6 mg/mL | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| DNAJB1 | |
| Full length human recombinant protein of human DNAJB1 (NP_006136) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1F9 | |
| Unconjugated | |
| DnaJ (Hsp40) homolog, subfamily B, member 1, DnaJ (Hsp40) homolog, subfmaily B, member 1, dnaJ homolog subfamily B member 1, DnaJ protein homolog 1, DNAJ1, hDj-1, HDJ1, Heat shock 40 kDa protein 1, heat shock 40kD protein 1, Heat shock protein 40, HSP40, HSPF1Hdj1, Human DnaJ protein 1, radial spoke 16 homolog B, RSPH16B, Sis1 | |
| Mouse | |
| 37.9 kDa | |
| 0.1 mL | |
| DNA Repair, Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
| 3337 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction