Learn More
Invitrogen™ HSP27 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1017
Description
PA1-017 detects heat shock protein 27(hsp27) from rat, mouse, monkey, and human tissue tissues. PA1-017 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, and immunohistochemistry procedures. By Western blot, this antibody detects a ~27 kDa protein which corresponds to hsp27 from HeLa cells. Immunocytochemical staining of hsp27 in HeLa cells with this antibody results in diffuse cytoplasmic staining before heat shock and diffuse cytoplasmic along with punctate staining patterns after. The PA1-017 immunogen is a synthetic peptide corresponding to residues L(10) L R G P S W D P F R C(21) of human Heat Shock Protein 27. This peptide is 81% conserved in mice, rats, and hamsters. The phosphorylated peptide (Cat. # PEP-187) and non-phosphorylated peptide (Cat. # PEP-240) are available for use in neutralization and control experiments.
In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock or stress proteins. HSPB1 (heat shock protein beta-1 or HSP27) is a small heat shock protein which functions as a molecular chaperone that maintains denatured proteins in a folding-competent state. It plays a role in stress resistance and actin organization. Through its molecular chaperone activity, HSP27 regulates numerous biological processes including the phosphorylation and the axonal transport of neurofilament proteins. Mutations in the gene can result in Charcot-Marie-Tooth disease 2F and Neuronopathy distal hereditary motor 2B.
Specifications
| HSP27 | |
| Polyclonal | |
| Unconjugated | |
| HSPB1 | |
| 27kDa; 28 kDa heat shock protein; CMT2F; DKFZp586P1322; epididymis secretory protein Li 102; estrogen-regulated 24 kDa protein; Growth-related 25 kDa protein; heat shock 25 kDa protein; Heat shock 27 kDa protein; heat shock 27kD protein 1; heat shock 27kDa protein 1; Heat shock protein; heat shock protein 1; heat shock protein 27 kDa beta-1; heat shock protein B1; heat shock protein beta-1; heat shock protein family B (small) member 1; heat shock protein, 25 kDa; heat-shock protein; HEL-S-102; HMN2B; HS; HS.76067; HSP; HSP 25; HSP 27; Hsp25; HSP27; HSP28; HSPB1; p25; Phospho-HSP 27; SRP27; Stress-responsive protein 27; truncated hsp25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 24471, 3315 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| P04792, P42930 | |
| HSPB1 | |
| Synthetic peptide corresponding to residues L(10) L R G P S W D P F R C(21) of human Heat Shock Protein 27. | |
| 100 μg | |
| Primary | |
| Human, Rat, Monkey | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.