missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16813-1-AP-20UL
This item is not returnable.
View return policy
Description
HSPB6 is associated with actin and modulates smooth muscle relaxation.Specifications
| HSP20 | |
| Polyclonal | |
| Unconjugated | |
| HSPB6 | |
| Heat shock protein beta 6, Hsp20, HSPB6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 126393, 192245, 243912 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14558, P97541, Q5EBG6 | |
| Hspb6 | |
| HSP20 Fusion Protein Ag10352 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction