missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HSP105 Recombinant Rabbit Monoclonal Antibody (23GB6190), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | HSP105 |
|---|---|
| Clone | 23GB6190 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088462
![]() |
Invitrogen™
MA553946 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087449
|
Invitrogen™
MA553947 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSP105 Recombinant Monoclonal antibody specifically detects HSP105 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HSP105 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q61699, Q66HA8, Q92598 | |
| 10808, 15505, 288444 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6190 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 105kDa; 42 degrees C-HSP; AI790491; antigen NY-CO-25; heat shock 105/110 protein 1; heat shock 105kD alpha; heat shock 105kD beta; heat shock 105kDa; heat shock 105kDa/110kDa protein 1; heat shock 110 kDa protein; Heat shock protein; Heat shock protein 105 kDa; heat shock protein 110; heat shock protein family H (Hsp110) member 1; heat shock protein, 105 kDa; heat shock-related 100 kDa protein E7I; heat-shock protein 110 kDa; HSP; Hsp105; HSP105 42 C-HSP; HSP105A; HSP105B; Hsp110; hsp110/105; HSP-E7I; Hsph1; I79_002075; Kiaa0201; KIAA0202; NY CO 25; NY-CO-25; SEP2; SEPT8; septin 8; SEPTIN8; septin-8 | |
| A synthesized peptide derived from human Hsp105 (51-99AA). | |
| Primary | |
| -20°C | |
| HSPH1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title